Merck
  • Phytoremediation potential of charophytes: bioaccumulation and toxicity studies of cadmium, lead and zinc.
Phytoremediation potential of charophytes: bioaccumulation and toxicity studies of cadmium, lead and zinc.
Journal of environmental sciences (China) (2013-08-08)
Najjapak Sooksawat, Metha Meetam, Maleeya Kruatrachue, Prayad Pokethitiyook, Koravisd Nathalang
ABSTRACT
The ability for usage of common freshwater charophytes, Chara aculeolata and Nitella opaca in removal of cadmium (Cd), lead (Pb) and zinc (Zn) from wastewater was examined. C. aculeolata and N. opaca were exposed to various concentrations of Cd (0.25 and 0.5 mg/L), Pb (5 and 10 mg/L) and Zn (5 and 10 mg/L) solutions under hydroponic conditions for 6 days. C. aculeolata was more tolerant of Cd and Pb than N. opaca. The relative growth rate of N. opaca was drastically reduced at high concentrations of Cd and Pb although both were tolerant of Zn. Both macroalgae showed a reduction in chloroplast, chlorophyll and carotenoid content after Cd and Pb exposure, while Zn exposure had little effects. The bioaccumulation of both Cd and Pb was higher in N. opaca (1544.3 microg/g at 0.5 mg/L Cd, 21657.0 microg/g at 10 mg/L Pb) whereas higher Zn accumulation was observed in C. aculeolata (6703.5 microg/g at 10 mg/L Zn). In addition, high bioconcentration factor values (> 1000) for Cd and Pb were observed in both species. C. aculeolata showed higher percentage of Cd and Pb removal (> 95%) than N. opaca and seemed to be a better choice for Cd and Pb removal from wastewater due to its tolerance to these metals.
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis