Merck
  • Effect of strontium ranelate on serum leptin and bone turnover markers in women with established postmenopausal osteoporosis.
Effect of strontium ranelate on serum leptin and bone turnover markers in women with established postmenopausal osteoporosis.
The Journal of reproductive medicine (2013-08-21)
Ilhan Bayhan, Nasuh U Dogan, Gulnur Ozaksit, Dilek Uygur, Nil Ugurlu, Mustafa Ugur
ABSTRACT
To assess the association between leptin levels and bone mineral density (BMD) and bone metabolism markers in postmenopausal women taking strontium ranelate. In this longitudinal uncontrolled study, postmenopausal osteoporotic women were administered strontium ranelate and categorized into 1 of 2 groups: overweight and lean. BMD, serum leptin, bone-specific alkaline phosphatase (ALP) and urinary deoxypyridinoline (Dpd) were assessed at baseline, 6 and 12 months of the therapy. For all 47 women, leptin levels increased at the sixth month (p = 0.001) and 1 year after treatment (p = 0.001), Dpd levels decreased at the sixth month (p = 0.001) and 1 year after treatment (p = 0.003), but ALP levels did not change with respect to basal levels. Spine and hip BMD also increased at the end of 1 year (p = 0.01 and p = 0.001, respectively). For overweight women leptin levels increased 6 months and 1 year after treatment (p = 0.003 and p = 0.001, respectively). Dpd levels decreased significantly at the sixth month and at 1 year (p = 0.001 and p = 0.041), but ALP levels did not change. Hip and spine BMD also increased (p = 0.036 and p = 0.002). The lean group had similar serum results and BMD scores after treatment. Our data supports a positive effect of leptin on bone metabolism in favor of bone resorption inhibition in postmenopausal osteoporotic women on strontium treatment.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein