Merck
Disposition and metabolism of 1-nitropropane in rats and chimpanzees.
Biomedical and environmental sciences : BES (1989-09-01)
M Haas-Jobelius, F Korte, F Coulston
ABSTRACT
The metabolic fate of 1-nitropropane (1-NP) has not been previously reported. In this study male rats and chimpanzees were given single doses of 40 mg/kg ip and 5 mg/kg iv 1-[1-14C]NP, respectively. The quantitative extent of urinary and fecal elimination was similar in both species. The rats excreted 16.5% of the dose in urine and 1.7% in feces. For chimpanzees the respective values were 14.8 and 1.2%. Experiments with rats demonstrated that the major route of elimination was by exhalation. With a total elimination via the lungs of 72.6%, rats expired 10.3% of the dose as unchanged 1-NP. Five polar metabolites were isolated from the urine of chimpanzees. The two major metabolites were identified as 3-hydroxypropionic acid and N-methyl-N-2-(methylsulfinyl)ethylpropionic acid amide (NMPA). Both substances were also excreted in rat urine. The two identified metabolites indicate that 1-NP was degraded to propionic acid, part of which was modified to 3-hydroxypropionic acid or NMPA. A hypothetical pathway for the biochemical generation of NMPA is suggested.
Sigma-Aldrich
1-Nitropropane, ≥98.5%