Merck
Identification of puberty-accelerating pheromones in male mouse urine.
The Journal of experimental zoology (1989-09-01)
K Nishimura, K Utsumi, M Yuhara, Y Fujitani, A Iritani
ABSTRACT
Gas chromatography-mass spectrometry was employed to identify the two volatile amines in male mouse urine. These amines were much less concentrated in urine of castrated males. The identified amines, isobutylamine and isoamylamine, were assayed for the potential of puberty acceleration in postweaning female mice. A total of 105 young female mice were exposed to one of the following five odors: distilled water (control), 0.1 M isobutylamine, 0.1 M isoamylamine, a mixture of 0.05 M isobutylamine and 0.05 M isoamylamine, or fresh male mouse urine. The mixture of these amines accelerated the vaginal opening of young females. Except for the control, all experimental odors accelerated the first vaginal estrus in ICR strain mice.
Sigma-Aldrich
1-Amino-3-methylbutane hydrochloride, puriss., ≥98.0% (TLC)
Sigma-Aldrich
Isopentylamine, ≥98%
Sigma-Aldrich
Isobutylamine, 99%
Sigma-Aldrich
Isopentylamine, 99%