Merck
  • Desorption chemical ionization and fast atom bombardment mass spectrometric studies of the glucuronide metabolites of doxylamine.
Desorption chemical ionization and fast atom bombardment mass spectrometric studies of the glucuronide metabolites of doxylamine.
Biomedical & environmental mass spectrometry (1986-11-01)
J O Lay, W A Korfmacher, D W Miller, P Siitonen, C L Holder, A B Gosnell
ABSTRACT
Three glucuronide metabolites of doxylamine succinate were collected in a single fraction using high-performance liquid chromatography (HPLC) from the urine of dosed male Fischer 344 rats. The metabolites were then separated using an additional HPLC step into fractions containing predominantly a single glucuronide metabolite. Analysis of the metabolites by methane and ammonia desorption chemical ionization, with and without derivatization, revealed fragment ions suggestive of a hydroxylated doxylamine moiety. Identification of the metabolites as glucuronides of doxylamine, desmethyldoxylamine and didesmethyldoxylamine was accomplished, based on determination of the molecular weight and exact mass of each metabolite using fast atom bombardment (FAB) ionization. This assignment was confirmed by the fragmentation observed in FAB mass spectrometric and tandem mass spectrometric experiments. Para-substitution of the glucuronide on the phenyl moiety was observed by 500-MHz nuclear magnetic resonance (NMR) spectrometry. A fraction containing all three glucuronide metabolites, after a single stage of HPLC separation, was also analysed by FAB mass spectrometry, and the proton- and potassium-containing quasimolecular ions for all three metabolites were observed.
Product Description
Supelco
Doxylamine succinate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Doxylamine succinate salt