Merck
  • Hydroxy acid metabolites of branched-chain amino acids in amniotic fluid.
Hydroxy acid metabolites of branched-chain amino acids in amniotic fluid.
Clinica chimica acta; international journal of clinical chemistry (1984-07-16)
C Jakobs, L Sweetman, W L Nyhan
ABSTRACT
A method is described in which ammonia chemical ionization gas chromatography-mass spectrometry was utilized in the selected ion monitoring mode to provide an accurate, selective approach to the quantification in amniotic fluid of a number of hydroxylated organic acids derived from the metabolism of the branched-chain amino acids. 2-Hydroxy-n-caproic acid was employed as an internal standard and the hydroxy acids were isolated from amniotic fluid by liquid partition chromatography and the trimethylsilyl derivatives were quantified. Normal values have been obtained for 2-hydroxyisovaleric acid, the sum of 2-hydroxyisocaproic acid and 2-hydroxy-3-methylvaleric acid, 2-methyl-3-hydroxybutyric acid, 3-hydroxyisovaleric acid and 2-ethyl-3-hydroxypropionic acid. The method also provides data on the concentration of methylmalonic acid. The concentration of 2-hydroxyisovaleric acid was not useful in the prenatal diagnosis of a fetus with maple syrup urine disease. Elevated concentrations of 2-methyl-3-hydroxybutyric acid as well as methylmalonic acid were found in the amniotic fluid of two fetuses with methylmalonic acidemia.
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%
Sigma-Aldrich
2-Hydroxyisocaproic acid, 99%
Sigma-Aldrich
D-α-Hydroxyisovaleric acid, ≥98.0% (T)
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
(S)-(+)-2-Hydroxy-3-methylbutyric acid, 99%, optical purity ee: 99% (GLC)
Sigma-Aldrich
(S)-(−)-2-Hydroxyisocaproic acid, 98%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG