Merck
  • The application of sodium deoxycholate and Sephacryl-200 for the delipidation and separation of high density lipoprotein.
The application of sodium deoxycholate and Sephacryl-200 for the delipidation and separation of high density lipoprotein.
Experientia (1982-04-15)
H Robern
ABSTRACT
A method to remove lipids from human plasma high-density lipoprotein (HDL) was developed. The procedure required column chromatography on Sephacryl-200 in the presence of the bile salt sodium deoxycholate. The lipid free protein obtained retained the immunological properties of the native HDL.
Sigma-Aldrich
Sephacryl®, 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 400-HR, MW range 20-8000 kDa (globular proteins)
Sigma-Aldrich
Sephacryl®, 100-HR, MW range 1000-100,000 Da (globular proteins)
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)