Merck
  • Modified trichrome staining technique of the nerve to determine proximal nerve viability.
Modified trichrome staining technique of the nerve to determine proximal nerve viability.
Microsurgery (1995-01-01)
R Meyer, G C Claussen, S J Oh
ABSTRACT
Normal myelinated fibers are stained "red" by modified trichrome staining of the frozen section of the nerve. We have used this technique for rapid assessment of the anatomical integrity of the proximal nerve stump by identifying well-preserved myelinated fibers. This technique can also identify degenerating and degenerated myelinated fibers, fibrosis, and inflammatory cells. We report three cases in which the practical usefulness of this technique is demonstrated.
Product Description
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %
Sigma-Aldrich
Fast Green FCF, certified by the BSC
Sigma-Aldrich
Chromotrope 2R, suitable for modified Gomori Trichrome stain