Merck
  • Conformational transformations in the protein lattice of phage P22 procapsids.
Conformational transformations in the protein lattice of phage P22 procapsids.
Biophysical journal (1993-07-01)
M L Galisteo, J King
ABSTRACT
During the packaging of double-stranded DNA by bacterial viruses, the precursor procapsid loses its internal core of scaffolding protein and undergoes a substantial expansion to form the mature virion. Here we show that upon heating, purified P22 procapsids release their scaffolding protein subunits, and the coat protein lattice expands in the absence of any other cellular or viral components. Following these processes by differential scanning calorimetry revealed four different transitions that correlated with structural transitions in the coat protein shells. Exit of scaffolding protein from the procapsid occurred reversibly and just above physiological temperature. Expansion of the procapsid lattice, which was exothermic, occurred after the release of scaffolding protein. Partial denaturation of coat subunits within the intact shell structure was detected prior to the major endothermic event. This major endotherm occurred above 80 degrees C and represents particle breakage and irreversible coat protein denaturation. The results indicate that the coat subunits are designed to form a metastable precursor lattice, which appears to be separated from the mature lattice by a kinetic barrier.
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 400-HR, MW range 20-8000 kDa (globular proteins)
Sigma-Aldrich
Sephacryl®, 100-HR, MW range 1000-100,000 Da (globular proteins)
Sigma-Aldrich
Sephacryl®, 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans)
Sigma-Aldrich
Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)