Merck
  • Application of stripping voltammetry to trace lead analysis in intermediates and final products of syntheses of pharmaceuticals.
Application of stripping voltammetry to trace lead analysis in intermediates and final products of syntheses of pharmaceuticals.
Journal of pharmaceutical and biomedical analysis (1996-05-01)
M Pravda, K Vytras
ABSTRACT
Applications of differential pulse anodic stripping voltammetry using a new pen-type renewed hanging mercury electrode have been investigated for trace analysis of lead in pharmaceutical substances and intermediates of their syntheses, such as procaine hydrochloride, 4-aminobenzoic acid, methyl 4-aminobenzoate, 2-(4-chlor-3-aminobenzoyl) benzoic acid, benzyl 2-naphthyl ether, 5-aminoisophthalic acid, 3-aminobenzoic acid, 5-hydroxyisophthalic acid, and N, N'-dibenzylethylenediamine diacetate. Samples were dissolved in 1 M HCI or 1 M NaOH and the electrochemical scan was carried out. No sample mineralization was necessary. The method showed a good linearity up to 50-100 ppm Pb with a detection limit less than 100 ppb. The results agreed well, but were more precise than those obtained by atomic absorption spectrometry using air/acetylene flame atomisation.
Sigma-Aldrich
5-Hydroxyisophthalic acid, 97%
Sigma-Aldrich
3-Aminobenzoic acid, 98%
Sigma-Aldrich
5-Aminoisophthalic acid, 94%
Sigma-Aldrich
Methyl 4-aminobenzoate, 98%