Merck
  • A new method to assign immunodetected spots in the complex two-dimensional electrophoresis pattern.
A new method to assign immunodetected spots in the complex two-dimensional electrophoresis pattern.
Electrophoresis (1997-05-01)
E Zeindl-Eberhart, P R Jungblut, H M Rabes
ABSTRACT
A new, easy method for the immunodetection of specific antigens in two-dimensional electrophoresis (2-DE) is described. Areas of 2-DE gels containing antigens of interest are electrophoretically transferred to polyvinylidene difluoride membranes, immunostained with specific antibodies using Fast Red or 5-bromo-4-chloro-3-indolyl phosphate/nitro blue tetrazolium as detection systems and counterstained with Coomassie Brilliant Blue. In contrast to conventional methods, it is possible to use this procedure to exactly assign immunoreacting proteins on a single blot to their corresponding and surrounding blue-stained protein spots.
Sigma-Aldrich
Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)
Sigma-Aldrich
Brilliant Blue R, pure
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution
Sigma-Aldrich
Brilliant Blue R, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade