Merck
  • The induction effect of rifampicin on activity of mephenytoin 4'-hydroxylase related to M1 mutation of CYP2C19 and gene dose.
The induction effect of rifampicin on activity of mephenytoin 4'-hydroxylase related to M1 mutation of CYP2C19 and gene dose.
British journal of clinical pharmacology (1998-03-07)
H J Feng, S L Huang, W Wang, H H Zhou
ABSTRACT
To determine the induction effect of rifampicin on the activity of 4'-hydroxylase in poor metabolizers (PMs) with m1 mutation of S-mephenytoin 4'-hydroxylation and the relationship of the effect with gene dose. Seven extensive metabolizers (EMs) of S-mephenytoin 4'-hydroxylation and five PMs with m1 mutation were chosen to take rifampicin 300 mg day(-1) orally for 22 days. Prior to and after rifampicin treatment, each subject was given racemic mephenytoin 100 mg. The 4'-hydroxymephenytoin (4'-OH-MP) excreted in the 0-24 h urine and mephenytoin S/R ratio in the 0-8 h urine were determined by h.p.l.c. and GC, respectively. In all EMs, the excretion of 4'-OH-MP in the 0-24 h urine was increased by 146.4 +/- 17.9%, 0-8 h urinary mephenytoin S/R ratio was decreased by 77.3 +/- 8.8%, the percentage increase in the 0-24 h excretion of 4'-OH-MP in those CYP2C19 homozygous (wt/wt) was greater than that in those heterozygous (wt/m1 and wt/m2) (203.9 +/- 42.5% vs 69.6 +/- 4.1%). 0-8 h urinary mephenytoin S/R ratio of those PMs with m1 mutation was decreased by 9.6%, the amount of 4'-OH-MP excreted in the 0-24 h urine was increased by 80.1 +/- 48.0%. The activity of 4'-hydroxylase of PMs with m1 mutation of S-mephenytoin 4'-hydroxylation can be induced by rifampicin and the inducing effect of rifampicin on 4'-hydroxylase is gene dependent.
Sigma-Aldrich
4-Hydroxymephenytoin
Sigma-Aldrich
4-Hydroxymephenytoin, ≥98% (HPLC)