Merck
  • [Effect on the microbicidal efficacy of formaldehyde, glutardialdehyde, peracetic acid, chloramine T (N-chloro-4-toluenesulfonamide), m-cresol, ethanol and benzyldimethyldodecacylammonium bromide by blood (model experiments for chemical disinfection of instruments)].
[Effect on the microbicidal efficacy of formaldehyde, glutardialdehyde, peracetic acid, chloramine T (N-chloro-4-toluenesulfonamide), m-cresol, ethanol and benzyldimethyldodecacylammonium bromide by blood (model experiments for chemical disinfection of instruments)].
Zentralblatt fur Hygiene und Umweltmedizin = International journal of hygiene and environmental medicine (1998-04-09)
G Spicher, J Peters
ABSTRACT
In a preceding paper (Zbl. Hyg. 191 [1991] 457-477) we reported on the dependence of the microbicidal efficacy of active agents of the disinfection of instruments on the amount of coagulated blood adhering to the instruments. In the present investigation, we were interested in the dependence of the microbicidal effects on the amount of blood in the solutions of the active agents. Test areas of 2 cm2 were contaminated with 50 and 100 microliters coagulating blood, respectively, containing cells of Staphylococcus aureus as test germ. The solutions of the microbicidal agents were contaminated with heparinized blood up to a concentration of 4% immediately before starting the disinfection and 24 hours before, respectively. After a period of action lasting 1 hour at 20 degrees C, the relative number of test germs capable of multiplying (N/N0) was determined. The concentration of the microbicidal substances reducing the relative number of test germs capable to multiply to 10(-4) served for estimating the dependence of the microbicidal efficacy of the agents on the blood content of the solutions. The experimental results depended on the thickness of the layer of coagulated blood. The dependence of the efficacy of the microbicidal substances on the blood content of the solutions was the higher the thinner the blood layer was. At a thickness of the layer of the coagulated blood of 0.25 mm, a blood content of the solution of 4%, and applying it immediately after adding the blood, the concentration of glutardialdehyde had to be 1.6 times that without blood to reach the same microbicidal efficacy. When applying the solution 24 hours after adding the blood, the concentration of glutardialdehyde had to be 4.2 times that without blood. The quaternary ammonium compound reacted faster with the blood than did glutardialdehyde; the respective factors were 2.6 and 4.5. The concentration factors of chloramine T were 3.3 and 3.8. Under the conditions of the test, peracetic acid exhibited small concentration factors: 1.3 and 1.6. The microbicidal efficacy of ethanol, formaldehyde and m-cresol soap solution was not or only slightly altered by the amount of blood in the solution of the microbicidal agent.
Sigma-Aldrich
Benzyldodecyldimethylammonium bromide, ≥99.0% (AT)
Sigma-Aldrich
Benzyldodecyldimethylammonium bromide
Sigma-Aldrich
Benzyldimethyldodecylammonium chloride, ≥99.0% (AT)