Merck
Clozapine--a dangerous drug in a clozapine-naïve subject.
Forensic science international (2011-08-13)
D Stanworth, N C A Hunt, R J Flanagan
ABSTRACT
Clozapine is a uniquely effective antipsychotic, but is very toxic in clozapine-naïve subjects. A 34-year-old male patient in a mental health facility, who was not prescribed clozapine, took 350 mg clozapine obtained from another patient at night. He was found dead the next morning. The presence of cardiomegaly related to obesity may have increased the risk of suffering an acute cardiac event after ingestion of clozapine. The medication prescribed to the patient was not thought to have contributed to the fatal outcome. Post mortem femoral blood clozapine and norclozapine concentrations were 0.48 and 0.20mg/L, respectively. By way of comparison, audit of 104,127 plasma samples (26,796 patients) assayed for therapeutic drug monitoring purposes 1993-2007, showed plasma clozapine 0.35 mg/L or more in 57.5% samples (8.4% 1mg/L or more). Those involved in the investigation of clozapine-associated deaths need to be aware that that death in an adult may occur after a single 'therapeutic' dose. A diagnosis of fatal clozapine poisoning cannot be made solely on the basis of a post mortem blood clozapine measurement.
Product Description
Supelco
N-Desmethylclozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
N-Desmethylclozapine