Merck
Effect of protective atmosphere on color of goose meat.
Poultry science (2013-07-23)
A Orkusz, J Woloszyn, G Haraf, A Okruszek
ABSTRACT
The objective of the work was to characterize the color of the of the goose breast meat packaged in protective atmosphere and stored in the refrigerated conditions. The aim was realized by determination of total heme pigment concentration; relative concentration of myoglobin, oxymyoglobin, and metmyoglobin; parameters of color L* (lightness), a* (redness), and b* (yellowness); and sensory evaluation of the surface color. The experimental material was White Kołuda goose boneless breast meat with the skin from industrial slaughter. The following 2 protective atmospheres were used in the study: vacuum and modified atmosphere (MA) consisting of 80% O2 and 20% CO2. The muscles packed in protective atmosphere were examined on d 4, 7, 11, and 14 of storage. A control sample was goose breast meat stored in air and tested after 24 h after slaughter. The total pigment concentration decreased gradually within 14 d of storage for samples packed in 2 types of atmospheres. The increase in relative concentration of metmyoglobin and the decrease in oxymyoglobin relative concentration in total heme pigments in the meat stored in MA was noticed. However, in all times of storage, the relative concentration of the 3 samples of myoglobin forms stored in vacuum was unchanged. The color parameters (L*, a*, b*) did not change for 14 d of storage in the muscles packed in vacuum. One can state a decrease of the value of the color parameter a* as well as an increase of the value of the color parameter b* in the samples packed in MA. From d 11 to 14 of storage, goose meat packed under MA had lower sensory evaluation intensity of color than muscles under vacuum. The obtained data indicated that the surface color of goose breast meat packed in MA (consisting of 80% O2, 20% CO2) or vacuum packed was maintained for 11 and 14 d, respectively.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C