Merck
  • Enhancing the intensities of lysine-terminated tryptic peptide ions in matrix-assisted laser desorption/ionization mass spectrometry.
Enhancing the intensities of lysine-terminated tryptic peptide ions in matrix-assisted laser desorption/ionization mass spectrometry.
Rapid communications in mass spectrometry : RCM (2000-12-13)
R L Beardsley, J A Karty, J P Reilly
ABSTRACT
Tryptic digests of three proteins are reacted with O-methylisourea in order to convert lysine residues to homoarginines. The resulting homoarginine-terminated peptides exhibit more intense MALDI mass spectral peaks than their lysine-terminated predecessors. This simple chemical reaction should therefore facilitate protein sequencing and mass mapping.
Sigma-Aldrich
O-Methylisourea hemisulfate salt, 99%
Sigma-Aldrich
o-Methylisourea bisulfate, 99%