Merck
The human PER1 gene is inducible by interleukin-6.
Journal of molecular neuroscience : MN (2002-04-05)
Dirk Motzkus, Urs Albrecht, Erik Maronde
ABSTRACT
The mammalian period (Per) genes which are components of the circadian clock are mainly regulated via an autoregulatory feedback loop. Here we show that a human PER1 (hPER1) reporter gene activity is stimulated by interleukin-6 (IL-6), a member of the large cytokine gene family and an inducer of the acute phase reaction, in human hepatoma (HuH-7) cells. Our results confirm and extend the view that the hPER1 promoter acts as a sensor for multiple signaling molecules thereby integrating different physiological parameters.