Merck
  • Impairment of testicular function in adult male Japanese quail (Coturnix japonica) after a single administration of 3-methyl-4-nitrophenol in diesel exhaust particles.
Impairment of testicular function in adult male Japanese quail (Coturnix japonica) after a single administration of 3-methyl-4-nitrophenol in diesel exhaust particles.
The Journal of endocrinology (2006-05-30)
ChunMei Li, Shinji Takahashi, Shinji Taneda, Chie Furuta, Gen Watanabe, Akira K Suzuki, Kazuyoshi Taya
ABSTRACT
The effects of 3-methyl-4-nitrophenol (PNMC), a component of diesel exhaust, on reproductive function were investigated in adult male Japanese quail. The quail were treated with a single i.m. dose of PNMC (78, 103 or 135 mg/kg body weight), and trunk blood and testes were collected 1, 2 or 4 weeks later. Various levels of testicular atrophy were observed in all groups treated with PNMC. Sperm formation, cloacal gland area, and plasma LH and testosterone concentrations were also reduced in birds with testicular atrophy. To determine the acute effect of PNMC on gonadotrophin from the pituitary, adult male quail were administrated a single i.m. injection of PNMC (25 mg/kg), and plasma concentrations of LH were measured at 1, 3 and 6 h. This dose significantly lowered plasma levels of LH at all three time points. These results suggest that PNMC acts on the hypothalamus-pituitary axis, by reducing circulating LH within a few hours of administration and subsequently reducing testosterone secretion. In addition, in order to investigate the direct effects of PNMC on the secretion of testosterone from testicular cells in quail testes, cultured interstitial cells containing Leydig cells were exposed to PNMC (10(-6), 10(-5) or 10(-4) M) for 4, 8 or 24 h. These quantities of PNMC significantly reduced the secretion of testosterone in a time- and dose-dependent manner. The present findings also suggest a direct effect of PNMC on the testis to reduce testosterone secretion. This study clearly indicates that PNMC induces reproductive toxicity at both the central and testicular levels, and disrupts testicular function in adult male quail.