Merck
Assessment of oritavancin serum protein binding across species.
Antimicrobial agents and chemotherapy (2010-05-26)
Francis F Arhin, Adam Belley, Geoffrey McKay, Sylvain Beaulieu, Ingrid Sarmiento, Thomas R Parr, Gregory Moeck
ABSTRACT
Biophysical methods to study the binding of oritavancin, a lipoglycopeptide, to serum protein are confounded by nonspecific drug adsorption to labware surfaces. We assessed oritavancin binding to serum from mouse, rat, dog, and human by a microbiological growth-based method under conditions that allow near-quantitative drug recovery. Protein binding was similar across species, ranging from 81.9% in human serum to 87.1% in dog serum. These estimates support the translation of oritavancin exposure from nonclinical studies to humans.