Merck
  • Successful treatment of carbamazepine poisoning with hemodialysis: a case report and review of the literature.
Successful treatment of carbamazepine poisoning with hemodialysis: a case report and review of the literature.
Hemodialysis international. International Symposium on Home Hemodialysis (2011-06-01)
Murugesan Ram Prabahar, Kalidindi Raja Karthik, Mohini Singh, Raja Beem Singh, Sudhagar Singh, Jayachandran Dhamodharan
ABSTRACT
A 48-year-old man was brought to the emergency room after ingesting an unknown amount of carbamazepine. He was unconscious and not responding to the noxious stimuli. He was intubated and was placed on mechanical ventilation because of respiratory insufficiency. Primary detoxification was performed with a gastric lavage and charcoal instillation. His serum carbamazepine level was 25.6 mcg/mL at the time of admission. His computed tomography of the brain was normal. He was managed conservatively but there was no improvement in his neurological status in the next 24 hours. Serum carbamazepine level was repeated and reported to be 28.3 mcg/mL. As there was no improvement in his sensorium and the serum carbamazepine levels remained persistently high, extracorporeal removal of carbamazepine was attempted. As the facility to carry out hemoperfusion was not available immediately, the decision to initiate hemodialysis was taken. After 3 sessions of hemodialysis, his sensorium improved markedly and the carbamazepine level at this time was within the therapeutic range. He was discharged after psychiatry consultation and counseling. We review the literature regarding extra corporeal techniques for the removal of carbamazepine and discuss them in this article.
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Carbamazepine 10,11-epoxide, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Carbamazepine-10,11-epoxide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®