Merck
  • Determination of the fatty acid profile of neutral lipids, free fatty acids and phospholipids in human plasma.
Determination of the fatty acid profile of neutral lipids, free fatty acids and phospholipids in human plasma.
Clinical chemistry and laboratory medicine (2012-09-26)
Nina Firl, Hermine Kienberger, Teresa Hauser, Michael Rychlik
ABSTRACT
Knowledge of the fatty acid composition of lipid classes in human plasma is an important factor in the investigation of human metabolism. Therefore, a method for the analysis of neutral lipid (NL), phospholipid (PL) and free fatty acids (FFA) in human plasma has been developed and validated. Separation of lipid classes was carried out by solid phase extraction of the lipid extract. The fractions were transesterified and the resulting fatty acid methyl esters were determined by GC/FID. For the method to be validated, precision, detection and quantification limits, as well as recovery, were determined for combined lipid extraction, solid phase extraction and GC analysis. The lipid extraction was miniaturized and simplified by application of an ultrasound ‘Sonotrode’. The resolution of lipid classes was optimized with appropriate standards added to a representative plasma sample. In addition, a rapid derivatization procedure using trimethylsulfoniumhydroxide was established. Low determination limits (1.5, 0.2 and 1.3 μg/g plasma for NL, PL and FFA, respectively) indicate that the method’s sensitivity is sufficient to quantify even minor components. Furthermore, recovery for NL and PL fatty acids was found to range from 80% to 110%. The results were similar for FFA apart from more polar free fatty acids due to their higher solubility in water. Repetitive measurements showed very good precision apart from the long chain PUFA for which the coefficients of variation were significantly higher. The present method is applicable to the quantitation of fatty acids in lipid classes of human plasma including several minor components.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.