Merck
Biochemical markers of fatal hypothermia.
Forensic science international (2013-01-15)
Cristian Palmiere, Daniel Bardy, Igor Letovanec, Patrice Mangin, Marc Augsburger, Francesco Ventura, Katia Iglesias, Dominique Werner
ABSTRACT
The aim of this study was to investigate the usefulness of postmortem biochemical investigations in the diagnosis of fatal hypothermia. 10 cases of fatal hypothermia and 30 control cases were selected. A series of biochemical parameters, such as glucose, acetone, 3-beta-hydroxybutyrate, isopropyl alcohol, free fatty acids, adrenaline, growth hormone, adrenocorticotropic hormone, thyroid-stimulating hormone, cortisol, calcium, magnesium, C-reactive protein, procalcitonin as well as markers of renal and cardiac functions were measured in blood, postmortem serum from femoral blood, urine, vitreous and pericardial fluid. The results suggested that deaths due to hypothermia, especially in free-ethanol cases, are characterized by increased ketone levels in blood and other biological fluids, increased adrenaline concentrations in urine, increased cortisol levels in postmortem serum from femoral blood and increased free cortisol values in urine. Increased or decreased levels of other biological parameters are either the result of terminal metabolic changes or the expression of preexisting diseases and may provide information to elucidate the death process on a case-by-case basis.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Isopropanol, 70% in H2O
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material