Merck
Modeling toxic stress by atrazine in a marine consumer-resource system.
Environmental toxicology and chemistry (2013-02-22)
Lisette De Hoop, Marleen De Troch, A Jan Hendriks, Frederik De Laender
ABSTRACT
The present study combines short-term experiments with food chain modeling to explore the long-term effects of the herbicide atrazine on consumer-resource dynamics in a marine intertidal ecosystem. Short-term (28 d) lab experiments indicated that the intrinsic rate of increase (r) and carrying capacity (K) of the marine diatom Seminavis robusta decreased with increasing atrazine exposure. This decrease did not show the concave shape expected from the lifetime productivity for nonexposed diatoms and from single-species toxicity data in the literature but instead was described best by a linear model. These experimentally observed atrazine-induced decreases of r and K were used to parameterize a Rosenzweig-MacArthur model representing a simple food chain including the tested diatom and its grazer, the harpacticoid copepod Delavalia palustris var. palustris. Stable oscillation zoo-phytobenthos systems were produced at diatom exposures of 0, 100, and 150 µg/L atrazine. An atrazine concentration of 150 µg/L contributed to a 15% increase of the oscillation periods of both diatoms and copepods as well as a 52% reduction of oscillation amplitudes compared with the control situation. Although the amplitudes of copepods increased only 7% at 150 µg/L atrazine, the maximum and minimum copepod densities at that concentration were reduced 61 and 63%, respectively. The effects of atrazine on periodicity and amplitudes were robust to 20% changes in the food-chain model parameters that represented allometric relationships. The simulations in the present study suggest food chain-mediated indirect effects on zoobenthos populations, indicating a reduced diatom and copepod availability throughout the year.
Product Description
Supelco
Atrazine, PESTANAL®, analytical standard
Supelco
Atrazine, analytical standard
Supelco
Atrazine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland