Merck
  • Effect of fatty acids on the β-oxidation system and thioesterase of Lactococcus lactis subspecies lactis.
Effect of fatty acids on the β-oxidation system and thioesterase of Lactococcus lactis subspecies lactis.
Journal of dairy science (2013-03-07)
Liang Li, Ying Ma
ABSTRACT
The influence of fatty acids on the β-oxidation system and thioesterase of Lactococcus lactis was investigated in this study. The results showed that fatty acids (C8:0-C16:0) significantly inhibited the growth of Lactococcus lactis, and laurate (C12:0) had the highest bactericidal effects. We detected the maximum activity of β-oxidation at different incubation times (8, 12, and 18h) to be 6.460, 7.751, and 8.203, respectively, and the maximum activity of thioesterase at different incubation times (8, 12, and 18h) to be 19.498, 27.180, and 12.800, respectively. Fatty acids were seen to induce the β-oxidation system and activity of thioesterase; decanoic acid (C10:0) and palmitic acid (C16:0) were also seen to induce the β-oxidation system of Lactococcus lactis, but the induced ability was significantly different. Octanoic acid (C8:0) and palmitic acid (C16:0) were seen to induce thioesterase activity in Lactococcus lactis. When 1mM palmitic acid (C16:0) was added to M17 broth, the activity of thioesterase increased 5-fold after 2min; however, adding octanoic acid (C8:0) changed the activity little. Evidence showed that the ability to induce the β-oxidation system and thioesterase activity was related to the fatty acids' chain lengths.
Product Description
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, natural, 98%, FG