Merck
  • Simultaneous analysis of guaiacol and vanillin in a vanilla extract by using high-performance liquid chromatography with electrochemical detection.
Simultaneous analysis of guaiacol and vanillin in a vanilla extract by using high-performance liquid chromatography with electrochemical detection.
Bioscience, biotechnology, and biochemistry (2013-03-09)
Makoto Takahashi, Shizuka Sakamaki, Akira Fujita
ABSTRACT
We developed and validated a new high-performance liquid chromatographic analysis for electrochemically detecting guaiacol and vanillin as important components in vanilla extract. Separation was achieved with Capcell Pak C-18 MG, the potential of the working electrode being set at +1000 mV. The respective calibration curves for guaiacol and vanillin were linear in the range of 1.60-460 µg/L and 5.90-1180 µg/L. The respective limits for the quantities of guaiacol and vanillin were 1.60 µg/L and 2.36 µg/L. The related standard deviations of the intra- and inter-day precision of the retention time and peak area were all less than 4%. The recovery of guaiacol and vanillin was both more than 97%, all of the validation data being within an acceptable range. This analysis method is well suited for the simultaneous and convenient analysis of guaiacol and vanillin in a vanilla extract to evaluate the quality of the vanilla extract.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG