Merck
  • Aegle marmelos fruit pectin for food and pharmaceuticals: Physico-chemical, rheological and functional performance.
Aegle marmelos fruit pectin for food and pharmaceuticals: Physico-chemical, rheological and functional performance.
Carbohydrate polymers (2013-03-19)
Manish Jindal, Vineet Kumar, Vikas Rana, A K Tiwary
ABSTRACT
Pectin is used in a number of foods as a gelling agent, thickener, texturizer, emulsifier and stabilizer. Bael fruit, obtained from Aegle marmelos, is a rich source of pectin. Bael fruit pectin (BFP) was extracted from ripe Bael fruits. The process yielded 15% (w/w) pure BFP. The swelling index decreased in the following order: water>pH 7.4>pH 6.8>pH 1.2>HCl (0.1N). Galacturonic acid content of 87.8%, degree of esterification of 47.2%, 17.3% methoxy groups, 0.29% acetyl groups and equivalent weight of 1209.5, indicate it to be a good gelling agent and easily amenable to derivatization. BFP exhibited a significant concentration-dependent prolongation of prothrombin time. The absence of hemagglutinating activity and antinutritional factors coupled with the activity to confer better emulsion capacity, stability and antimicrobial activity gives BFP a clear edge over commercial citrus pectin (CP) for exploitation as an additive in food and pharmaceuticals.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
D-Galacturonic acid sodium salt, ≥95.0% (T)