Merck
  • Biocompatible nanostructured magnesium oxide-chitosan platform for genosensing application.
Biocompatible nanostructured magnesium oxide-chitosan platform for genosensing application.
Biosensors & bioelectronics (2013-03-19)
Manoj Kumar Patel, Md Azahar Ali, Md Zafaryab, Ved Varun Agrawal, M Moshahid Alam Rizvi, Z A Ansari, S G Ansari, Bansi D Malhotra
ABSTRACT
A novel organic-inorganic platform comprising of chitosan (CH) modified nanostructured magnesium oxide (nanoMgO) has been electrophoretically deposited on the indium-tin-oxide (ITO) substrate. The single stranded probe DNA (ssDNA) sequence of Vibrio cholerae has been covalently functionalized onto CH-nanoMgO/ITO surface. The cytotoxicity assay of nanoMgO particles, examined using human intestinal cell line (INT 407), reveals no significant cytotoxicity at the given doses in the range of 50-350 μg/mL. The X-ray diffraction (XRD), Fourier transform infrared spectroscopy (FT-IR) and various microscopic techniques have been employed for the structural and morphological analysis of the fabricated electrodes. The electrochemical response studies of ssDNA and fragmented genomic DNA hybridized electrode (dsGDNA/CH-nanoMgO/ITO) have been carried out using cyclic voltammetry (CV) and differential pulse voltammetry (DPV) techniques. The dsGDNA/CH-nanoMgO/ITO bioelectrode exhibits a linear response in the range 100-500 ng/μL with improved sensitivity of 36.72 nA/ng/cm(2), faster response time of 3s and high stability of 3-4 months under refrigerated condition. The lower detection limit of fabricated genosensor has been estimated as 35.20 ng/μL and it shows good reproducibility/repeatability.
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis