Merck
  • Risk factors for neutropenia in clozapine-treated children and adolescents with childhood-onset schizophrenia.
Risk factors for neutropenia in clozapine-treated children and adolescents with childhood-onset schizophrenia.
Journal of child and adolescent psychopharmacology (2013-03-21)
Kristin N Maher, Marcus Tan, Julia W Tossell, Brian Weisinger, Peter Gochman, Rachel Miller, Deanna Greenstein, Gerald P Overman, Judith L Rapoport, Nitin Gogtay
ABSTRACT
The purpose of this study was to retrospectively analyze rates of neutropenia and risk factors for neutropenia in hospitalized children and adolescents treated with clozapine. A retrospective chart review was conducted for all patients who received clozapine at any time during a hospitalization at the National Institute of Mental Health (NIMH) between 1990 and 2011. All patients satisfied screening criteria for the NIMH childhood-onset schizophrenia study, including onset of psychosis before the age of 13 years. Absolute neutrophil count (ANC) values recorded during inpatient hospitalization were extracted for 87 eligible patients with a mean age of 13.35±2.46 years at hospitalization and a mean length of stay of 117±43 days. Mild neutropenia only (lowest ANC<2000/mm3 but>1500/mm3) was observed in 27 (31%) patients and moderate neutropenia (any ANC<1500/mm3) was observed in 17 (20%) patients. There were no cases of agranulocytosis or severe infection. Significant risk factors for mild neutropenia compared with no hematologic adverse effects (HAEs) were male gender (p=0.012) and younger age (p<0.001). Male gender was also a significant risk factor for moderate neutropenia compared with no HAEs (p=0.003). If a child of African American ethnicity developed neutropenia during hospitalization at all that child was significantly more likely to develop moderate neutropenia than mild neutropenia only (p=0.017). African American boys had the highest rate of moderate neutropenia at 47%. Sixteen of the 17 patients exhibiting moderate neutropenia were successfully treated with clozapine by the time of discharge; 8 of these 16 required adjunctive lithium carbonate administration to maintain ANC>2000/mm3. Our study shows that the rates of neutropenia in clozapine-treated children and adolescents are considerably higher than in the adult population. Younger age, African American ethnicity, and male gender were significant risk factors. These are also risk factors for benign neutropenia in healthy children and adolescents. Despite these high rates of neutropenia, all but one of the patients with neutropenia during hospitalization were successfully discharged on clozapine.
Product Description
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clozapine
Clozapine, European Pharmacopoeia (EP) Reference Standard