Merck
  • A prospective 10-year study of metal ceramic single crowns and fixed dental prosthesis retainers in private practice settings.
A prospective 10-year study of metal ceramic single crowns and fixed dental prosthesis retainers in private practice settings.
The Journal of prosthetic dentistry (2013-03-26)
Bernd Reitemeier, Kristina Hänsel, Christian Kastner, Anke Weber, Michael H Walter
ABSTRACT
Metal ceramic restorations are widely used in prosthodontics, but long-term data on their clinical performance in private practice settings based on prospective trials are sparse. This clinical trial was designed to provide realistic long-term survival rates for different outcomes related to tooth loss, crown loss, and metal ceramic defect. Ninety-five participants were provided with 190 noble metal ceramic single crowns and 138 participants with 276 fixed dental prosthesis retainer crowns on vital posterior teeth. Follow-up examinations were scheduled 2 weeks after insertion, annually up to 8 years, and after 10 years. Kaplan-Meier survival analyses, Mantel-Cox logrank tests, and Cox regression analyses were conducted. Because of variations in the time of the last examinations, the maximum observation period was 12.1 years. For the primary outcome 'loss of crown or tooth', the Kaplan-Meier survival rate was 94.3% ±1.8% (standard error) at 8.0 years (last outcome event) for single crowns and 94.4% ±1.5% at 11.0 years for fixed dental prosthesis retainer crowns. The difference between the survival functions was not significant (P>.05). For the secondary outcome 'metal ceramic defect', the survival rate was 88.8% ±3.2% at 11.0 years for single crowns and 81.7% ±3.5% at 11.0 years for fixed dental prosthesis retainer crowns. In Cox regression models, the only significant covariates for the outcome event 'metal ceramic defect' were bruxism in the medical history (single crowns) and signs and symptoms of bruxism (fixed dental prosthesis retainer crowns) with hazard ratios of 3.065 (95% CI 1.063 - 8.832) and 2.554 (95% CI 1.307 - 4.992). Metal ceramic crowns provided in private practice settings show good longevity. Bruxism appears to indicate a risk for metal ceramic defects.
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, sponge, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade