Merck
  • Influence of citric acid and curing on moisture sorption, diffusion and permeability of starch films.
Influence of citric acid and curing on moisture sorption, diffusion and permeability of starch films.
Carbohydrate polymers (2013-04-03)
Erik Olsson, Mikael S Hedenqvist, Caisa Johansson, Lars Järnström
ABSTRACT
Starch films with different amounts of citric acid produced by solution casting were subjected to different curing temperatures and compared with films plasticized with glycerol. The films were tested in a controlled moisture generator, which enabled the moisture sorption to be measured and the diffusion coefficient and water vapor permeability to be calculated. It was shown that increasing the amount of citric acid added led to a reduction in the equilibrium moisture content, diffusion coefficient and water vapor permeability of the films, the values of which were all considerably lower than the values obtained for the films plasticized by glycerol. It was also seen that curing the film with 30 pph citric acid at 150°C led to a significant reduction in the equilibrium moisture content, the diffusion coefficient and the water vapor permeability at high relative humidity which suggests that crosslinking occurred. The calculated water vapor permeability data were comparable with the value obtained with direct measurements.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, Powder
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Starch from rice
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)