Merck
  • Topical amitriptyline combined with ketamine for the treatment of erythromelalgia: a retrospective study of 36 patients at Mayo Clinic.
Topical amitriptyline combined with ketamine for the treatment of erythromelalgia: a retrospective study of 36 patients at Mayo Clinic.
Journal of drugs in dermatology : JDD (2013-04-03)
Timothy J Poterucha, William T Weiss, Roger A Warndahl, Richard H Rho, Paola Sandroni, Mark D P Davis, Sinead L Murphy
ABSTRACT
Erythromelalgia is an uncommon neurovascular disorder characterized by redness, increased skin temperature, and pain that usually occurs in the extremities. Treatment remains challenging because of its varying response to medical therapy. The objective of this study was to assess the response of erythromelalgia to compounded topical amitriptyline-ketamine. We retrospectively evaluated 36 patients with erythromelalgia who were treated with compounded topical amitriptyline-ketamine from January 1, 2004, through January 31, 2011. Thirty-two patients (89%) were female. Mean (standard deviation) age was 44.7 (15.8) years (range, 5-74 years). Patients applied the medication 1 to 6 times per day (median, 5 times). One patient (3%) had complete relief from symptoms, 14 (39%) had substantial relief, 12 (33%) had some relief, 7 (19%) had no relief, and 2 (6%) had local worsening of symptoms. No patients had systemic adverse effects. A majority of patients with erythromelalgia (75%) reported improvement in pain with topical application of a compounded amitriptyline-ketamine formulation. The medication was well tolerated.
Product Description
Supelco
Ketamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®