Merck
  • Toward an ultra-broadband emission source based on the bismuth and erbium co-doped optical fiber and a single 830nm laser diode pump.
Toward an ultra-broadband emission source based on the bismuth and erbium co-doped optical fiber and a single 830nm laser diode pump.
Optics express (2013-04-03)
Jianzhong Zhang, Zinat M Sathi, Yanhua Luo, John Canning, Gang-Ding Peng
ABSTRACT
We demonstrate a broadband optical emission from Bi/Er co-doped fiber and a single 830nm laser diode pump. The ultra-broadband mechanism is studied and discussed in details based on a combination of experimental measurements, including luminescence, differential luminescence and ESA, on fiber samples of different Bi and Er concentrations. The Er co-doping in Bi doped fiber is found to be effective for broadband emission, by enhancing not only luminescence at C and L bands but also that at O and shorter wavelength bands. The luminescence intensity between 1100 and 1570nm is over -45dBm/5nm in single mode fiber using a few meters of Bi/Er co-doped fiber and offers a modest ~40dB dynamic range and a broad bandwidth of ~470nm for an OSA based spectral measurement.
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis