Merck
  • Acid-soluble internal capsules for closed-face cassette elemental sampling and analysis of workplace air.
Acid-soluble internal capsules for closed-face cassette elemental sampling and analysis of workplace air.
Journal of occupational and environmental hygiene (2013-04-04)
Martin Harper, Kevin Ashley
ABSTRACT
Airborne particles that are collected using closed-face filter cassettes (CFCs), which are used widely in the sampling of workplace aerosols, can deposit in places other than on the filter and thereby may not be included in the ensuing analysis. A technique for ensuring that internal non-filter deposits are included in the analysis is to collect airborne particles within an acid-soluble internal capsule that, following sampling, can be dissolved along with the filter for subsequent elemental analysis. An interlaboratory study (ILS) was carried out to evaluate the use of cellulosic CFC capsule inserts for their suitability in the determination of trace elements in airborne samples. The ILS was performed in accordance with an applicable ASTM International standard practice, ASTM E691, which describes statistical procedures for investigating interlaboratory precision. Performance evaluation materials consisted of prototype cellulose acetate capsules attached to mixed-cellulose ester filters. Batches of capsules were dosed with Pb-containing materials (standard aqueous solutions, and certified reference material soil and paint). Also, aerosol samples containing nine target analyte elements (As, Cd, Co, Cr, Cu, Fe, Pb, Mn, and Ni) were generated using a multiport sampler; various concentrations and sampling times were employed to yield samples fortified at desired loading levels. Triplicates of spiked capsules at three different loadings were conveyed to each volunteer laboratory; loading levels were unknown to the participants. The laboratories were asked to prepare the samples by acid dissolution and to analyze aliquots of extracted samples by atomic spectrometry in accordance with applicable ASTM International Standards. Participants were asked to report their results in units of μg of each target element per sample. For the elements investigated, inter-laboratory precision and recovery estimates from the participating laboratories demonstrated the utility of the cellulosic capsule inserts for the measurement of sampled trace elements.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)