Merck
  • Plasmalemmal sodium-calcium exchanger shapes the calcium and exocytotic signals of chromaffin cells at physiological temperature.
Plasmalemmal sodium-calcium exchanger shapes the calcium and exocytotic signals of chromaffin cells at physiological temperature.
American journal of physiology. Cell physiology (2013-04-19)
Juan-Fernando Padín, José-Carlos Fernández-Morales, Román Olivares, Stefan Vestring, Juan-Alberto Arranz-Tagarro, Enrique Calvo-Gallardo, Ricardo de Pascual, Luís Gandía, Antonio G García
ABSTRACT
The activity of the plasmalemmal Na(+)/Ca(2+) exchanger (NCX) is highly sensitive to temperature. We took advantage of this fact to explore here the effects of the NCX blocker KB-R7943 (KBR) at 22 and 37°C on the kinetics of Ca(2+) currents (ICa), cytosolic Ca(2+) ([Ca(2+)]c) transients, and catecholamine release from bovine chromaffin cells (BCCs) stimulated with high K(+), caffeine, or histamine. At 22°C, the effects of KBR on those parameters were meager or nil. However, at 37°C whereby the NCX is moving Ca(2+) at a rate fivefold higher than at 22°C, various of the effects of KBR were pronounced, namely: 1) no effects on ICa; 2) reduction of the [Ca(2+)]c transient amplitude and slowing down of its rate of clearance; 3) blockade of the K(+)-elicited quantal release of catecholamine; 4) blockade of burst catecholamine release elicited by K(+); 5) no effect on catecholamine release elicited by short K(+) pulses (1-2 s) and blockade of the responses produced by longer K(+) pulses (3-5 s); and 6) potentiation of secretion elicited by histamine or caffeine. Furthermore, the more selective NCX blocker SEA0400 also potentiated the secretory responses to caffeine. The results suggest that at physiological temperature the NCX substantially contributes to shaping the kinetics of [Ca(2+)]c transients and the exocytotic responses elicited by Ca(2+) entry through Ca(2+) channels as well as by Ca(2+) release from the endoplasmic reticulum.
Product Description
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Histamine, ≥97.0%
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%