Merck
  • Multilevel functional and structural defects induced by two pathogenic mitochondrial tRNA mutations.
Multilevel functional and structural defects induced by two pathogenic mitochondrial tRNA mutations.
The Biochemical journal (2013-05-02)
Meng Wang, Xiao-Long Zhou, Ru-Juan Liu, Zhi-Peng Fang, Mi Zhou, Gilbert Eriani, En-Duo Wang
ABSTRACT
Point mutations in hmtRNAs (human mitochondrial tRNAs) can cause various disorders, such as CPEO (chronic progressive external ophthalmoplegia) and MM (mitochondrial myopathy). Mitochondrial tRNALeu, especially the UUR codon isoacceptor, is recognized as a hot spot for pathogenic mtDNA point mutations. Thus far, 40 mutations have been reported in hmtRNAsLeu. In the present paper, we describe the wide range of effects of two substitutions found in the TΨC arms of two hmtRNAsLeu isoacceptors. The G52A substitution, corresponding to the pathogenic G12315A mutation in tRNALeu(CUN), and G3283A in tRNALeu(UUR) exhibited structural changes in the outer corner of the tRNA shape as shown by RNase probing. These mutations also induced reductions in aminoacylation, 3'-end processing and base modification processes. The main effects of the A57G substitution, corresponding to mutations A12320G in tRNALeu(CUN) and A3288G in tRNALeu(UUR), were observed on the aminoacylation activity and binding to hmEF-Tu (human mitochondrial elongation factor Tu). These observations suggest that the wide range of effects may amplify the deleterious impact on mitochondrial protein synthesis in vivo. The findings also emphasize that an exact understanding of tRNA dysfunction is critical for the future development of therapies for mitochondrial diseases.
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder