Merck
  • Effect of epidural levobupivacaine and levobupivacaine with fentanyl on stress response and postoperative analgesia after total knee replacement.
Effect of epidural levobupivacaine and levobupivacaine with fentanyl on stress response and postoperative analgesia after total knee replacement.
International journal of clinical pharmacology and therapeutics (2013-06-21)
Esra Gumus Bayazit, Kazim Karaaslan, Kutay Ozturan, Erdinc Serin, Hasan Kocoglu
ABSTRACT
Providing sufficient and convenient analgesia is crucial during the postoperative period after totalknee replacement (TKR) to enhance patient mobility and reduce stress response to surgery. The scope of this study is to compare the effects of levobupivacaine and levobupivacaine plus fentanyl on stress response and analgesic efficiency after TKR. In this study, 40 ASA I - II patients scheduled to undergo TKR were subjected to combined spinal epidural anesthesia (CSEA) injecting of 15 mg levobupivacaine and randomly assigned to receive either levobupivacaine 0.125% (Group L) or levobupivacaine 0.125% plus fentanyl 4 μg ml-1 (Group F) during postoperative period via the epidural route. Patient controlled epidural analgesia (PCEA) was offered for 24 hours. Venous blood samples were assayed for adrenocorticotropic hormone (ACTH), cortisol and prolactin levels before surgery and after analgesia administration. Analgesia was assessed using a visual analogue scale (VAS) at rest (VASR) and during movement (VASM). There was no statistically significant difference between the groups in terms of total doses, bolus requests, bolus delivered and side effects (p > 0.05). The ACTH, cortisol and prolactin levels increased following the surgery and decreased during PCA infusion in both groups where the decline in Group F was significant (p < 0.05) at 24 hours after the analgesic treatment and 48 hours after the surgery. We have demonstrated that infusion of levobupivacaine (0.125%) in combination with fentanyl (4 μg ml-1) using PCEA suppressed stress response to surgery significantly and provided better pain relief than levobupivacaine (0.125%) alone after TKR.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Prolactin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®