Merck
Do patients on statins also need niacin?
JAAPA : official journal of the American Academy of Physician Assistants (2013-08-08)
Kelly W Jones
ABSTRACT
Niacin has been used for years to treat hypercholesterolemia, but may not be as beneficial as thought for patients at high risk of cardiovascular disease. Until further research is done, niacin should be used only in patients who do not tolerate or respond to statin therapy, and never in patients who have achieved goal LDL cholesterol on a statin.
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material