Merck
  • Neuroprotection by bilobalide in ischemia: improvement of mitochondrial function.
Neuroprotection by bilobalide in ischemia: improvement of mitochondrial function.
Die Pharmazie (2013-08-09)
T M Schwarzkopf, S Hagl, G P Eckert, J Klein
ABSTRACT
Bilobalide, an active constituent of Ginkgo biloba, is known to have neuroprotective properties, but its mode of action remains unclear. In this study, bilobalide significantly reduced brain damage in mice (indicated by TTC staining) when given before transient middle cerebral artery occlusion (tMCAO). As measured by microdialysis in the ischemic striatum, local perfusion with bilobalide (10 microM) reduced ischemia-induced glutamate release by 70% while glucose levels were not affected. Mitochondria isolated from ischemic brain showed a decrease of respiration compared to non-ischemic controls. Treatment with bilobalide (10 mg/kg) before tMCAO improved respiratory capacity of complex I significantly when measured ex vivo. In addition, mitochondrial swelling induced ex vivo by calcium was used to estimate opening of the mitochondrial permeability transition pore. In this assay, the changes induced by tMCAO were completely reversed when mice had received pretreatment with bilobalide. We conclude that neuroprotection by bilobalide involves a mechanism in which the drug reverses ischemia-induced changes in mitochondria, leading to a reduction of glutamate release.
Product Description
Supelco
(−)-Bilobalide, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
(−)-Bilobalide from Ginkgo biloba leaves, ≥93% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, FCC
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material