Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Aluminum fluoride, powder, 99.8% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host