Merck
  • Microsomal metabolism of pyrrolizidine alkaloids: N-oxidation of seneciphylline and senecionine.
Microsomal metabolism of pyrrolizidine alkaloids: N-oxidation of seneciphylline and senecionine.
Toxicology letters (1987-08-01)
H S Ramsdell, D R Buhler
ABSTRACT
In vivo pretreatment of rats with phenobarbital or beta-naphthoflavone reduced the specific activity of microsomal pyrrolizidine alkaloid N-oxide formation. Heat pretreatment of microsomes under conditions intended to selectively inactivate the flavin-containing monooxygenase did not lower the rate of N-oxidation. Incubation in the presence of cytochrome P-450 inhibitors diminished the microsomal formation of N-oxide. The observations are consistent with the hypothesis that pyrrolizidine alkaloid N-oxidation