- Effect of isoenzyme selective phosphodiesterase inhibitors on the proliferation of murine thymus and spleen cells.
Effect of isoenzyme selective phosphodiesterase inhibitors on the proliferation of murine thymus and spleen cells.
The effects of isoenzyme selective phosphodiesterase inhibitors on mitogen-stimulated proliferation of murine thymus and spleen cells was determined. The type 4 phosphodiesterase inhibitors, (rolipram, RO 20-1724 and denbufylline) and the mixed type 3/4 inhibitors, (zardaverine and benzafentrine) produced a concentration-related inhibition of mitogen stimulated thymus and spleen cell proliferation. Combined addition of the type 4 inhibitor, rolipram and the type 3 inhibitor, SK&F 94836 had no antiproliferative effect additional to that of rolipram alone. The thymus cells were more sensitive to the type 4 inhibitors than the spleen cells. The type 3 phosphodiesterase inhibitor, SK&F 94836 significantly inhibited cell proliferation, but only at high concentrations. The type 1 (vinpocetine) and the type 5 (zaprinast) phosphodiesterase inhibitor had no significant effect on proliferation. These results suggest that thymus and to a lesser extent spleen cell proliferation is dependent on the activity of the type 4 phosphodiesterase isoenzyme.