Merck
  • Biochemical synthesis, purification and preliminary pharmacological evaluation of normorphine-3-glucuronide.
Biochemical synthesis, purification and preliminary pharmacological evaluation of normorphine-3-glucuronide.
Life sciences (1997-01-01)
G D Smith, R J Prankerd, M T Smith
ABSTRACT
Normorphine was synthesised from morphine by thermal decomposition of an N-alpha-chloroethylchloroformate adduct, and purified (> 98% purity) using semi-preparative HPLC with ultraviolet detection. Normorphine-3-glucuronide (NM3G) was biochemically synthesised using the substrate normorphine, uridine diphosphoglucuronic acid and Sprague-Dawley rat liver microsomes in a 75% yield (relative to normorphine base). The synthesised NM3G was purified by precipitation and washing with acetonitrile. Determinations of purity using HPLC with electrochemical and ultraviolet detection confirmed that the NM3G produced was of high (> 99%) purity. Mass spectrometry, fourier transform infrared spectrophotometry and nuclear magnetic resonance spectrometry confirmed the structure, especially placement of the glucuronide moiety at the 3-phenolic position and not at the 17-nitrogen. Administration of NM3G by the intracerebroventricular (icv) route to rats in doses of 2.5 and 7.5 microg resulted in the development of central nervous system (CNS) excitatory behavioural effects including myoclonus, chewing, wet-dog shakes, ataxia and explosive motor behaviour. At an icv dose of 7.5 microg, NM3G also induced short periods of tonic-clonic convulsive activity. Thus, NM3G elicits CNS excitation following supraspinal administration in a manner analogous to morphine-3-glucuronide (M3G), the major metabolite of morphine (1). Further studies are required to determine whether NM3G attenuates morphine-induced antinociception in a similar manner to M3G.