Merck
  • Significance of L-alloisoleucine in plasma for diagnosis of maple syrup urine disease.
Significance of L-alloisoleucine in plasma for diagnosis of maple syrup urine disease.
Clinical chemistry (1999-10-03)
P Schadewaldt, A Bodner-Leidecker, H W Hammen, U Wendel
ABSTRACT
The significance of plasma L-alloisoleucine, which is derived from L-isoleucine in vivo, for diagnosis of maple syrup urine disease (MSUD) was examined. Branched-chain L-amino acids were measured by automatic amino acid analysis. Alloisoleucine reference values in plasma were established in healthy adults [1.9 +/- 0.6 micromol/L (mean +/- SD); n = 35], children 3-11 years (1.6 +/- 0.4 micromol/L; n = 17), and infants <3 years (1.3 +/- 0.5 micromol/L; n = 37). The effect of dietary isoleucine was assessed in oral loading tests. In controls receiving 38 micromol (n = 6; low dose) and 1527 micromol (n = 3; high dose) of L-isoleucine per kilogram of body weight, peak increases of plasma isoleucine were 78 +/- 24 and 1763 +/- 133 micromol/L, respectively; the peak increase of alloisoleucine, however, was negligible for low-dose (<0.3 micromol/L) and minor for high-dose (5. 5 +/- 2.1 micromol/L) load. In patients with diabetes mellitus, ketotic hypoglycemia, phenylketonuria, and obligate heterozygous parents of MSUD patients, alloisoleucine was not significantly different from healthy subjects. Therefore, a plasma concentration of 5 micromol/L was used as a cutoff value. In patients with classical MSUD (n = 7), alloisoleucine was beyond the cutoff value in 2451 of 2453 unselected samples. In patients with variant MSUD (n = 9), alloisoleucine was >5 micromol/L in all samples taken for establishment of diagnosis and in 94% of the samples taken for treatment control (n = 624). With the other branched-chain amino acids, the frequency of diagnostically significant increases was <45%. The present findings indicate that plasma L-alloisoleucine above the cutoff value of 5 micromol/L is the most specific and most sensitive diagnostic marker for all forms of MSUD.
Product Description
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Valine
Sigma-Aldrich
(S)-(−)-2-Hydroxyisocaproic acid, 98%
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, 99%, FG
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Isoleucine
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)