Merck
Cell death of dopamine neurons in aging and Parkinson's disease.
Mechanisms of ageing and development (2000-02-03)
M Naoi, W Maruyama
ABSTRACT
Dopamine neurons in the substantia nigra of human brain are selectively vulnerable and the number decline by aging at 5-10% per decade. Enzymatic and non-enzymatic oxidation of dopamine generates reactive oxygen species, which induces apoptotic cell death in dopamine neurons. Parkinson's disease (PD) is also caused by selective cell death of dopamine neurons in this brain region. The pathogenesis of Parkinson's disease remains to be an enigma, but it was found that an endogenous MPTP-like neurotoxin, 1(R), 2(N)-dimethyl-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline [N-methyl(R)salsolinol, NM(R)Sal], may be one of the pathogenic agents of PD. NM(R)Sal increases in cerebrospinal fluid from untreated parkinsonian patients, and two enzymes, a (R)salsolinol synthase and a neutral N-methyltransferase, synthesize this neurotoxin in the nigro-striatum. The activity of a neutral N-methyltransferase is significantly higher in lymphocytes from parkinsonian patients than in control. The mechanism of cell death by this toxin was proved to be by the induction of apoptosis, by use of dopaminergic SH-SY5Y cells. The apoptosis was suppressed by anti-oxidants, suggesting that the generation of reactive oxygen species may initiate cellular death process. These results indicate that in aging and PD oxidative stress induces degeneration of dopamine neurons, and the antioxidant therapy may delay the decline of dopamine neurons in the brain.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis