Merck
  • Simultaneous analysis of naproxen, nabumetone and its major metabolite 6-methoxy-2-naphthylacetic acid in pharmaceuticals and human urine by high-performance liquid chromatography.
Simultaneous analysis of naproxen, nabumetone and its major metabolite 6-methoxy-2-naphthylacetic acid in pharmaceuticals and human urine by high-performance liquid chromatography.
Journal of pharmaceutical and biomedical analysis (2000-10-07)
E Mikami, T Goto, T Ohno, H Matsumoto, M Nishida
ABSTRACT
A high-performance liquid chromatographic (HPLC) method for simultaneous determination of naproxen (NAP), nabumetone (NAB) and its major metabolite, 6-methoxy-2-naphthylacetic acid (6-MNA), was developed for the application to pharmaceuticals and human urine. Isocratic reversed-phase HPLC was employed for quantitative analysis using triethylamine and 1-heptanesulfonic acid sodium salt (HSA) as ion-pair reagents. Urine samples were purified by solid-phase extraction using Bond-Elut Certify II cartridges containing reversed-phase and anion exchange functionalities. The HPLC assay was carried out using a Wakosil ODS 5C18 column (5 microm, 150 x 4.6 mm, i.d.). The mobile phase consisted of 0.5 g of HSA dissolved in 1,000 ml of a mixture of acetonitrile, water and triethylamine (500:500:1, v/v) adjusted with phosphoric acid to pH 3. The calibration curves of NAP and NAB showed good linearity in the concentration range 32-160 microg/ml with UV detection (270 nm) for pharmaceuticals. In the low concentration ranges (8-96 ng of NAP per ml, 24-288 ng of NAB per ml and 5.6-67.2 ng of 6-MNA per ml), the calibration curves were also obtained with fluorimetric detection (excitation 280 nm, emission 350 nm) for biological fluids. The correlation coefficients were better than 0.999 in all cases. The lower limits of detection (defined as a signal-to-noise ratio of about 3) were approximately 0.3 ng for NAP, 1.5 ng for NAB and 0.2 ng for 6-MNA. The procedure described here is rapid, simple, selective, and is suitable for routine analysis of pharmaceuticals and pharmacokinetic studies in human urine samples.
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard