Merck
  • Metabolism of [(14)C]omapatrilat, a sulfhydryl-containing vasopeptidase inhibitor in humans.
Metabolism of [(14)C]omapatrilat, a sulfhydryl-containing vasopeptidase inhibitor in humans.
Drug metabolism and disposition: the biological fate of chemicals (2000-12-22)
R A Iyer, J Mitroka, B Malhotra, S Bonacorsi, S C Waller, J K Rinehart, V A Roongta, K Kripalani
ABSTRACT
Omapatrilat, a potent vasopeptidase inhibitor, is currently under development for the treatment of hypertension and congestive heart failure. This study describes the plasma profile along with isolation and identification of urinary metabolites of omapatrilat from subjects dosed orally with 50 mg of [(14)C]omapatrilat. Only a portion of the radioactivity in plasma was unextractable (40-43%). Prominent metabolites identified in plasma were S-methyl omapatrilat, acyl glucuronide of S-methyl omapatrilat, and S-methyl (S)-2-thio-3-phenylpropionic acid. Omapatrilat accounted for less than 3% of the radioactivity. However, after dithiothreitol reduction all of the radioactivity was extractable and was characterized to be omapatrilat and its hydrolysis product (S)-2-thio-3-phenylpropionic acid, both apparently bound to proteins via reversible disulfide bonds. Urinary profile of radioactivity showed no parent compound but the presence of several metabolites that can be grouped into three categories. 1) Three metabolites, accounting for 56% of the urinary radioactivity, resulted from the hydrolysis of the exocyclic amide bond of omapatrilat. Two metabolites were diastereomers of S-methyl sulfoxide of (S)-2-thio-3-phenylpropionic acid, and the third was the acyl glucuronide of S-methyl (S)-2-thio-3-phenylpropionic acid. 2) One disulfide, identified as the L-cysteine mixed disulfide of omapatrilat, accounted for 8% of the radioactivity in the urine. 3) Five metabolites, derived from omapatrilat, accounted for 30% of the radioactivity in the urine. Two of these metabolites were mixtures of diastereomers of S-methyl sulfoxide of omapatrilat and the third was the S-methyl omapatrilat ring sulfoxide. The other two metabolites were S-methyl omapatrilat and its acyl glucuronide. These results indicate that omapatrilat undergoes extensive metabolism in humans.
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
Hydrocinnamic acid, 99%