Merck
  • Sensitive and simple determination of mannitol in human brain tissues by gas chromatography-mass spectrometry.
Sensitive and simple determination of mannitol in human brain tissues by gas chromatography-mass spectrometry.
Journal of chromatography. B, Biomedical sciences and applications (2001-08-03)
A Kiyoshima, K Kudo, Y Hino, N Ikeda
ABSTRACT
A simple, reliable and sensitive gas chromatographic-mass spectrometric method was devised to determine the level of mannitol in various human brain tissues obtained at autopsy. Mannitol was extracted with 10% trichloroacetic acid solution which effectively precipitated brain tissues. The supernatant was washed with tert.-butyl methyl ether to remove other organic compounds and to neutralize the aqueous solution. Mannitol was then derivatized with 1-butaneboronic acid and subjected to GC-MS. Erythritol was used as an internal standard. For quantitation, selected ion monitoring with m/z 127 and 253 for mannitol and m/z 127 for internal standard were used. Calibration curves were linear in concentration range from 0.2 to 20 microg/0.1 g and correlation coefficients exceeded 0.99. The lower detection limit of mannitol in distilled water was 1 ng/0.1 g. Mannitol was detected in control brain tissues, as a biological compound, at a level of 50 ng/0.1 g. The precision of this method was examined with use of two different concentrations, 2 and 20 microg/0.1 g, and the relative standard deviation ranged from 0.8 to 8.3%. We used this method to determine mannitol in brain tissues from an autopsied individual who had been clinically diagnosed as being brain dead. Cardiac arrest occurred 4 days later.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture