Merck
  • Quantitative analysis of desmosterol, cholesterol and cholesterol sulfate in semen by high-performance liquid chromatography.
Quantitative analysis of desmosterol, cholesterol and cholesterol sulfate in semen by high-performance liquid chromatography.
Journal of chromatography. A (2002-01-05)
B Sion, G Grizard, D Boucher
ABSTRACT
A simple, rapid and accurate method to separate and quantify cholesterol, desmosterol and cholesterol sulfate in human spermatozoa and seminal plasma (SP) is described. This high-performance liquid chromatographic procedure is based on reversed-phase chromatography on a Inertsil ODS2 5 microm silica column with a binary gradient of mixtures of chloroform-methanol and chloroform-methanol-water as the mobile phase at a flow-rate of 0.25 ml/min. Sterols are separated with good resolution and high reproducibility. The eluted sterols are quantified using a light-scattering (mass) detector. As little as 64, 64 and 68 pmol of cholesterol, desmosterol and cholesterol sulfate, respectively, can be quantified under these conditions. Cholesterol is the predominant sterol both in spermatozoa (107+/-7 nmol/10(8) spermatozoa) and SP (0.83+/-0.10 micromol/ml) whereas the concentrations of desmosterol were 38+/-6 nmol/10(8) in spermatozoa and 0.18+/-0.02 micromol/ml in SP. Cholesterol sulfate represents about 6% of total cholesterol in the spermatozoa and SP. In conclusion, this method offers interesting perspectives for the quantitative analysis of these sterols not only in semen, but also in other biological samples.
Sigma-Aldrich
Desmosterol, ≥84% (GC)