Merck
  • Plasma hormones and catecholamine metabolites in monozygotic twins discordant for psychosis.
Plasma hormones and catecholamine metabolites in monozygotic twins discordant for psychosis.
Neuropsychiatry, neuropsychology, and behavioral neurology (2002-03-06)
Elaine F Walker, Robert Bonsall, Deborah J Walder
ABSTRACT
To examine plasma cortisol, adrencorticotropin hormone, and indicators of catecholamine activity in monozygotic twins discordant for schizophrenia or major affective disorders. Previous research has suggested that catecholamines play a role in the etiology of major mental illness. Several findings have also shown an inverse relation between hippocampal volume and cortisol levels in psychiatric populations including patients who are depressed and patients with Cushing disease. In this study, plasma obtained from monozygotic twin pairs discordant for schizophrenia (n = 10) or major affective disorder (n = 3) was assayed for epinephrine, norepinephrine, dihydroxyphenylacetic acid, homovanillic acid, adrencorticotropin hormone, and cortisol. There was significant concordance for levels of dihydroxyphenylacetic acid and homovanillic acid, consistent with the high concordance for indicators of dopamine activity observed in healthy monozygotic pairs. There was also concordance for adrencorticotropin hormone. However, in contrast to findings on healthy monozygotic pairs, there was no relation for epinephrine, norepinephrine, or cortisol. Among patients, there was an inverse correlation between cortisol and the magnitude of the reduction in hippocampal volume, relative to that of the healthy co-twin. These findings suggest the potential role of adrenal steroids and hippocampal function in the expression of psychosis.
Product Description
SAFC
L-Aspartic acid
SAFC
L-Tyrosine
SAFC
L-Threonine
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Tyrosine, FG
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)