Merck
  • Serum uric acid levels in multiple sclerosis patients correlate with activity of disease and blood-brain barrier dysfunction.
Serum uric acid levels in multiple sclerosis patients correlate with activity of disease and blood-brain barrier dysfunction.
European journal of neurology (2002-05-03)
G Toncev, B Milicic, S Toncev, G Samardzic
ABSTRACT
Several findings suggest lower levels of serum uric acid in multiple sclerosis (MS) patients. The aim of this study is to investigate relationships of uric acid serum levels in relapse-remitting (RR) MS patients with clinical activity of disease and blood-brain barrier (BBB) condition. Sixty-three definite RRMS patients and 40 controls divided into two groups: 20 healthy donors and 20 patients with other inflammatory neurological diseases (OINDs) were analysed. By using a quantitative enzymatic assay according to the manufacture's protocol and a commercial uric acid standard solution, serum uric acid levels were measured and the results were standardized. To investigate BBB function, magnetic resonance imaging after administration of gadolinium was used. MS patients were found to have significantly lower serum uric acid levels (193.89 +/- 49.05 micromol/l; mean value +/-SD) in comparison with healthy donors (292.7 +/- 58.65 micromol/l; P=0.000) and OIND patients (242.7 +/- 46.66 micromol/l; P=0.001). We found that MS patients with relapse had significantly lower serum uric acid levels (161.49 +/- 23.61 micromol/l) than MS patients with remission (234.39 +/- 41.96 micromol/l; P=0.000) and more over, MS patients with BBB disruption had significantly lower serum uric acid levels (163.95 +/- 26.07 micromol/l) than those with normal BBB (252.48 +/- 25.94 micromol/l; P=0.000). Further, we also found that serum uric acid level independently correlated with disease activity, BBB disruption, and gender. These results indicate that lower uric acid levels in MS patients are associated with relapse and suggest that uric acid might be beneficial in the treatment of MS.
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Coenzyme Q1, ≥95%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis